Learn More
Biomatik Corporation Mouse Fgf23 (Fibroblast growth factor 23) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKF57797
Encompass_Preferred
Mouse Fgf23 (Fibroblast growth factor 23) ELISA Kit for detecting Fgf23 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 15.625-1000pg/ml. Sensitivity: 9.375pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: Q9EPC2. Target Synonyms: FGF23; ADHR; FGF-23; fibroblast growth factor 23; HPDR2; HYPF; Phosphatonin; phosphatonin; PHPTC; tumor-derived hypophosphatemia inducing factor; Tumor-derived hypophosphatemia-inducing factor. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-0969