Learn More
Biomatik Corporation Mouse ACCPA (anti-cyclic citrullinated peptide antibody) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKF57803
Encompass_Preferred
Mouse ACCPA (anti-cyclic citrullinated peptide antibody) ELISA Kit for detecting ACCPA in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 1.563-100ng/ml. Sensitivity: 0.938ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antigen. Assay Time: 3h, 30 min. Target Synonyms: ACCPA; anti-cyclic citrullinated peptide antibody. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-0975