Learn More
Biomatik Corporation Mouse IGFBP-7 (Insulin-like growth factor-binding protein 7) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKF57812
Encompass_Preferred
Mouse IGFBP-7 (Insulin-like growth factor-binding protein 7) ELISA Kit for detecting IGFBP-7 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 78.125-5000pg/ml. Sensitivity: 46.875pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: Q61581. Target Synonyms: IGFBP-7; AGM; FSTL2; IBP-7; IGFBP7; IGFBP-7v; IGFBPRP1; MAC25; PSF; RAMSVPS; TAF. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-0984