Learn More
Biomatik Corporation Mouse A42 (Amyloid Beta 42) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKF57813
Encompass_Preferred
Mouse Aβ42 (Amyloid Beta 42) ELISA Kit for detecting Aβ42 in serum, plasma, cell culture supernatant, cell or tissue lysate, cerebrospinal fluid (csf), other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 15.625-1000pg/ml. Sensitivity: 9.375pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P12023. Target Synonyms: Aβ42; Amyloid Beta 42; Aβ; 1-42; Aβ1-42. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-0985