Learn More
Biomatik Corporation Mouse FSH (Follicle-stimulating hormone) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKF57815
Encompass_Preferred
Mouse FSH (Follicle-stimulating hormone) ELISA Kit for detecting FSH in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 2.344-150miu/ml. Sensitivity: 1.406miu/ml. Format: 96-Well Plate. Assay Type: Competitive ELISA, Coated with Antigen. Assay Time: 3h, 30 min. Accession Number: P35378. Target Synonyms: FSH; Follicle-Stimulating Hormone. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-0987