Learn More
Biomatik Corporation Mouse Hepc (Hepcidin) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKF57816
Encompass_Preferred
Mouse Hepc (Hepcidin) ELISA Kit for detecting Hepc in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 3.125-200ng/ml. Sensitivity: 1.875ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: Q9EQ21. Target Synonyms: Hepc; Hepcidin; HAMP; HFE2B; LEAP1; PLTR; LEAP-1; hepcidin antimicrobial peptide; Putative liver tumor regressor; Liver-expressed antimicrobial peptide 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-0988