Learn More
Biomatik Corporation Mouse LH (Luteinizing Hormone) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKF57817
Encompass_Preferred
Mouse LH (Luteinizing Hormone) ELISA Kit for detecting LH in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.469-30ng/ml. Sensitivity: 0.281ng/ml. Format: 96-Well Plate. Assay Type: Competitive ELISA, Coated with Antigen. Assay Time: 3h, 30 min. Accession Number: O09108; P01216. Target Synonyms: LH; Luteinizing Hormone; Lutropin; Lutrophin. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-0989