Learn More
Biomatik Corporation Mouse PAF (Platelet Activating Factor) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKF57822
Encompass_Preferred
Mouse PAF (Platelet Activating Factor) ELISA Kit for detecting PAF in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.094ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Target Synonyms: Platelet Activating Factor. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-0994