Learn More
Biomatik Corporation Mouse sIL-6R (Soluble Interleukin 6 Receptor) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKF57823
Encompass_Preferred
Mouse sIL-6R (Soluble Interleukin 6 Receptor) ELISA Kit for detecting sIL-6R in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 12.5-800pg/ml. Sensitivity: 7.5pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P22272. Target Synonyms: sIL-6R. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-0995