Learn More
Biomatik Corporation Rat Lipocalin-2/NGAL (Neutrophil Gelatinase Associated Lipocalin) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKF57832
Encompass_Preferred
Rat Lipocalin-2/NGAL (Neutrophil Gelatinase Associated Lipocalin) ELISA Kit for detecting Lipocalin-2; NGAL in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 62.5-4000pg/ml. Sensitivity: 37.5pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P30152. Target Synonyms: Lipocalin-2; LCN2; NGAL; Siderocalin; 24p3; 25 kDa alpha-2-microglobulin-related subunit of MMP-9; HNL; LCN2; lipocalin 2; lipocalin 2; oncogene 24p3; Lipocalin-2; migRation-stimulating factor inhibitor; MSFI; neutrophil gelatinase-associated lipocalin; NGALlipocalin-2; Oncogene 24p3; p25; siderocalin. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-1004