Learn More
Biomatik Corporation Rat BDNF (Brain Derived Neurotrophic Factor) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKF57834
Encompass_Preferred
Rat BDNF (Brain Derived Neurotrophic Factor) ELISA Kit for detecting BDNF in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 31.25-2000pg/ml. Sensitivity: 18.75pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P23363. Target Synonyms: BDNF; Brain Derived Neurotrophic Factor; ANON2; BULN2; Abrineurin; neurotrophin. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-1006