Learn More
Biomatik Corporation Rat BMP-2 (Bone Morphogenetic Protein 2) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKF57835
Encompass_Preferred
Rat BMP-2 (Bone Morphogenetic Protein 2) ELISA Kit for detecting BMP-2 in cell or tissue lysate, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 62.5-4000pg/ml. Sensitivity: 37.5pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P49001. Target Synonyms: BMP-2; Bone Morphogenetic Protein 2; BMP2A; BMP-2A. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-1007