Learn More
Biomatik Corporation Rat IP-10/CXCL10 (Interferon Gamma Induced Protein 10kDa) ELISA Kit, 96 tests
Catalog No. 501501017
Encompass_Preferred
Rat IP-10/CXCL10 (Interferon Gamma Induced Protein 10kDa) ELISA Kit for detecting IP-10; CXCL10 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 31.25-2000pg/ml. Sensitivity: 18.75pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P48973. Target Synonyms: CXCL10; IP-10; Interferon Gamma Induced Protein 10kDa; C7; IFI10; INP10; SCYB10; crg-2; gIP-10; mob-1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-1017 Supplier Biomatik Corporation Supplier No. EKF57845
Rat IP-10/CXCL10 (Interferon Gamma Induced Protein 10kDa) ELISA Kit for detecting IP-10; CXCL10 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 31.25-2000pg/ml. Sensitivity: 18.75pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P48973. Target Synonyms: CXCL10; IP-10; Interferon Gamma Induced Protein 10kDa; C7; IFI10; INP10; SCYB10; crg-2; gIP-10; mob-1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.