Learn More
Biomatik Corporation Rat IGF-1 (Insulin-Like Growth Factor 1) ELISA Kit, 96 tests
Catalog No. 501501018
Encompass_Preferred
Rat IGF-1 (Insulin-Like Growth Factor 1) ELISA Kit for detecting IGF-1 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 62.5-4000pg/ml. Sensitivity: 37.5pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P08025. Target Synonyms: IGF-1; Insulin-Like Growth Factor 1; IGF1; IGFI; IGF-I; IGF1A; IGF-IA; IGF-IB; MGF; Somatomedin C. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-1018 Supplier Biomatik Corporation Supplier No. EKF57846
Rat IGF-1 (Insulin-Like Growth Factor 1) ELISA Kit for detecting IGF-1 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 62.5-4000pg/ml. Sensitivity: 37.5pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P08025. Target Synonyms: IGF-1; Insulin-Like Growth Factor 1; IGF1; IGFI; IGF-I; IGF1A; IGF-IA; IGF-IB; MGF; Somatomedin C. Restrictions: For Research Use Only. Not for use in diagnostic procedures.