Learn More
Biomatik Corporation Rat IL-18 (Interleukin-18) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKF57851
Encompass_Preferred
Rat IL-18 (Interleukin-18) ELISA Kit for detecting IL-18 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 31.25-2000pg/ml. Sensitivity: 18.75pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P97636. Target Synonyms: IL-18; Interleukin 18; IL18; IGIF; IL-1g; IL-1F4; Interferon-gamma-inducing factor. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-1023