Learn More
Biomatik Corporation Rat IL-2 (Interleukin-2) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKF57852
Encompass_Preferred
Rat IL-2 (Interleukin-2) ELISA Kit for detecting IL-2 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 31.25-2000pg/ml. Sensitivity: 18.75pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P17108. Target Synonyms: IL-2; Interleukin 2; IL2; TCGF; Lymphokine. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-1024