Learn More
Biomatik Corporation Rat IL-3 (Interleukin-3) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKF57853
Encompass_Preferred
Rat IL-3 (Interleukin-3) ELISA Kit for detecting IL-3 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 15.625-1000pg/ml. Sensitivity: 9.375pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P04823. Target Synonyms: IL-3; IL3; MCGF; MCSF; MULTI-CSF; P-Cell Stimulating Factor. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-1025