Learn More
Biomatik Corporation Rat MCP-1 (Monocyte Chemotactic Protein 1) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKF57857
Encompass_Preferred
Rat MCP-1 (Monocyte Chemotactic Protein 1) ELISA Kit for detecting MCP-1 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 15.625-1000pg/ml. Sensitivity: 9.375pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P14844. Target Synonyms: MCP-1; CCL2; GDCF-2; HC11; HSMCR30; MCAF; MCP1; SCYA2; SMC-CF. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-1029