Learn More
Biomatik Corporation Rat NOS3/eNOS (Nitric Oxide Synthase 3, Endothelial) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKF57875
Encompass_Preferred
Rat NOS3/eNOS (Nitric Oxide Synthase 3, Endothelial) ELISA Kit for detecting NOS3; eNOS in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 15.625-1000pg/ml. Sensitivity: 9.375pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: Q62600. Target Synonyms: NOS3; eNOS; Constitutive NOS; ECNOS; endothelial; Endothelial NOS; eNOS; cNOS; nitric oxide synthase 3; endothelial cell. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-1037