Learn More
Biomatik Corporation Rat NOS2/iNOS (Nitric Oxide Synthase 2, Inducible) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKF57876
Encompass_Preferred
Rat NOS2/iNOS (Nitric Oxide Synthase 2, Inducible) ELISA Kit for detecting NOS2; iNOS in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.313-20ng/ml. Sensitivity: 0.188ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: Q06518. Target Synonyms: NOS2; iNOS; NOS2A; HEP-NOS; I-NOS; Hepatocyte NOS; Peptidyl-cysteine S-nitrosylase NOS2; Inducible NO synthase; Inducible NOS; INOS; NOS; NOS; type II; NOS type II. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-1038