Learn More
Biomatik Corporation Rat Stat3 (Signal transducer and activator of transcription 3) ELISA Kit, 96 tests
Catalog No. 501501041
Encompass_Preferred
Rat Stat3 (Signal transducer and activator of transcription 3) ELISA Kit for detecting Stat3 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.094ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P52631. Target Synonyms: Stat3; Signal transducer and activator of transcription 3. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-1041 Supplier Biomatik Corporation Supplier No. EKF57879
Rat Stat3 (Signal transducer and activator of transcription 3) ELISA Kit for detecting Stat3 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.094ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P52631. Target Synonyms: Stat3; Signal transducer and activator of transcription 3. Restrictions: For Research Use Only. Not for use in diagnostic procedures.