Learn More
Biomatik Corporation Rat Fst (Follistatin) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKF57881
Encompass_Preferred
Rat Fst (Follistatin) ELISA Kit for detecting Fst in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.094ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P21674. Target Synonyms: Fst; Follistatin. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-1043