Learn More
Biomatik Corporation Rat Ctgf (Connective tissue growth factor) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKF57885
Encompass_Preferred
Rat Ctgf (Connective tissue growth factor) ELISA Kit for detecting Ctgf in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 31.25-2000pg/ml. Sensitivity: 18.75pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: Q9R1E9. Target Synonyms: Ctgf; CCN2; HCS24; IGFBP-8; NOV2; CCN2IGFBP-8; HCS24; Hypertrophic chondrocyte-specific protein 24; IBP-8; IGF-binding protein 8; IGFBP8CCN family member 2; Insulin-like growth factor-binding protein 8; MGC102839. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-1047