Learn More
Biomatik Corporation Rat Pgf (Placenta growth factor) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKF57887
Encompass_Preferred
Rat Pgf (Placenta growth factor) ELISA Kit for detecting Pgf in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 7.813-500pg/ml. Sensitivity: 4.688pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: Q63434. Target Synonyms: Pgf; PGF; PGFL; D12S1900; PGFL; placenta growth factor; placental growth factor; placental growth factor; vascular endothelial growth factor-related protein; PlGF; PlGF-2; PLGFplacental growth factor-like; SHGC-10760;. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-1049