Learn More
Biomatik Corporation Rat Ahsg (Alpha-2-HS-glycoprotein) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKF57888
Encompass_Preferred
Rat Ahsg (Alpha-2-HS-glycoprotein) ELISA Kit for detecting Ahsg in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 78.125-5000pg/ml. Sensitivity: 46.875pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P24090. Target Synonyms: Ahsg; AHSG; alpha-2-HS-glycoprotein; A2HS; AHS; alpha-2-HS-glycoprotein; alpha-2-Z-globulin; Alpha-2-Z-globulin; Ba-alpha-2-glycoprotein; FETUAba-alpha-2-glycoprotein; fetuin-A; Fetuin A; HSGA. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-1050