Learn More
Biomatik Corporation Rat Cav-1 (Caveolin-1) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKF57890
Encompass_Preferred
Rat Cav-1 (Caveolin-1) ELISA Kit for detecting Cav-1 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 15.625-1000pg/ml. Sensitivity: 9.375pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P41350. Target Synonyms: Cav-1; CAV1; MSTP085; VIP21; CAV; caveolae protein; 22kD; caveolin 1; caveolae protein; 22kDa; cell growth-inhibiting protein 32; MSTP085. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-1052