Learn More
Biomatik Corporation Rat Crhr2 (Corticotropin-releasing factor receptor 2) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKF57892
Encompass_Preferred
Rat Crhr2 (Corticotropin-releasing factor receptor 2) ELISA Kit for detecting Crhr2 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.313-20ng/ml. Sensitivity: 0.188ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P47866. Target Synonyms: Crhr2; Corticotropin-releasing hormone receptor 2; CRH-R-2; CRH-R2;. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-1054