Learn More
Biomatik Corporation Rat Lpl (Lipoprotein lipase) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKF57898
Encompass_Preferred
Rat Lpl (Lipoprotein lipase) ELISA Kit for detecting Lpl in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.781-50ng/ml. Sensitivity: 0.469ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: Q06000. Target Synonyms: Lpl. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-1059