Learn More
Biomatik Corporation Rat Ghrh (Growth Hormone Releasing Hormone) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKF57904
Encompass_Preferred
Rat Ghrh (Growth Hormone Releasing Hormone) ELISA Kit for detecting Ghrh in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 15.625-1000pg/ml. Sensitivity: 9.375pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P09916. Target Synonyms: Ghrh; GRF; GHRF; Somatorelin; Somatocrinin; Growth hormone-releasing factor. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-1065