Learn More
Biomatik Corporation Rat NT-proBNP (N-Terminal Pro-Brain Natriuretic Peptide) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKF57906
Encompass_Preferred
Rat NT-proBNP (N-Terminal Pro-Brain Natriuretic Peptide) ELISA Kit for detecting NT-proBNP in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 31.25-2000pg/ml. Sensitivity: 18.75pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P13205. Target Synonyms: NT-proBNP; N-Terminal Pro-Brain Natriuretic Peptide. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-1067