Learn More
Biomatik Corporation Rat PICP (C-terminal propeptide of Collagen alpha-1 (I) chain) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKF57908
Encompass_Preferred
Rat PICP (C-terminal propeptide of Collagen alpha-1 (I) chain) ELISA Kit for detecting PICP in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 1.25-80ng/ml. Sensitivity: 0.75ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P02454; P02466. Target Synonyms: PICP; C-Propeptide Of Type I Procollagen; C-terminal propeptide of Collagen alpha-1; I chain. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-1069