Learn More
Biomatik Corporation Rat PIIINP (N-terminal propeptide of Collagen alpha-1 (III) chain) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKF57909
Encompass_Preferred
Rat PIIINP (N-terminal propeptide of procollagen type III) ELISA Kit for detecting PIIINP in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.313-20ng/ml. Sensitivity: 0.188ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P13941. Target Synonyms: PIIINP; P3NP; N-Propeptide Of Type III Procollagen; Procollagen III Amino Terminal Propeptide. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-1070