Learn More
Biomatik Corporation Rat Mif (Macrophage migration inhibitory factor) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKF57914
Encompass_Preferred
Rat Mif (Macrophage migRation inhibitory factor) ELISA Kit for detecting Mif in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.094ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P30904. Target Synonyms: Mif; Macrophage migRation inhibitory factor; GIFmacrophage migRation inhibitory factor; GLIF; Glycosylation-inhibiting factor; L-dopachrome isomerase; L-dopachrome tautomerase; macrophage migRation inhibitory factor; glycosylation-inhibiting factor; MMIF; Phenylpyruvate tautomerase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-1075