Learn More
Biomatik Corporation Rat Ttr (Transthyretin) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKF57917
Encompass_Preferred
Rat Ttr (Transthyretin) ELISA Kit for detecting Ttr in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 3.125-2000ng/ml. Sensitivity: 18.75ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P02767. Target Synonyms: Ttr; PALB; TBPA; ATTR; Prealbumin; HsT2651; prealbumin; amyloidosis type I; thyroxine-binding prealbumin. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-1078