Learn More
Biomatik Corporation Rat Iapp (Islet amyloid polypeptide) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKF57920
Encompass_Preferred
Rat Iapp (Islet amyloid polypeptide) ELISA Kit for detecting Iapp in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 78.125-5000pg/ml. Sensitivity: 46.875pg/ml. Format: 96-Well Plate. Assay Type: Competitive ELISA, Coated with Antigen. Assay Time: 3h, 30 min. Accession Number: P12969. Target Synonyms: Iapp; IAPP; DAP; IAP; Amylin; Islet amyloid polypeptide; Insulinoma amyloid peptide; Diabetes-associated peptide; DAP. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-1081