Learn More
Biomatik Corporation Rat CK-MB (Creatine Kinase MB Isoenzyme) ELISA Kit, 96 tests
Catalog No. 501501087
Encompass_Preferred
Rat CK-MB (Creatine Kinase MB Isoenzyme) ELISA Kit for detecting CK-MB in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 31.25-2000pg/ml. Sensitivity: 18.75pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P07335. Target Synonyms: CK-MB. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-1087 Supplier Biomatik Corporation Supplier No. EKF57926
Rat CK-MB (Creatine Kinase MB Isoenzyme) ELISA Kit for detecting CK-MB in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 31.25-2000pg/ml. Sensitivity: 18.75pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P07335. Target Synonyms: CK-MB. Restrictions: For Research Use Only. Not for use in diagnostic procedures.