Learn More
Biomatik Corporation Rat CTXI (Cross Linked C-telopeptide of Type I Collagen) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKF57929
Encompass_Preferred
Rat CTXI (Cross Linked C-telopeptide of Type I Collagen) ELISA Kit for detecting CTXI in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.094ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P02454; P02466. Target Synonyms: CTXI; CTX-I. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-1090