Learn More
Biomatik Corporation Rat GnRH (Gonadotropin-Releasing Hormone) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKF57934
Encompass_Preferred
Rat GnRH (Gonadotropin-Releasing Hormone) ELISA Kit for detecting GnRH in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 15.625-1000pg/ml. Sensitivity: 9.375pg/ml. Format: 96-Well Plate. Assay Type: Competitive ELISA, Coated with Antigen. Assay Time: 3h, 30 min. Accession Number: P07490. Target Synonyms: GnRH; Gonadotropin Releasing Hormone. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-1094