Learn More
Biomatik Corporation Rat HbA1C (Glycosylated Hemoglobin/Hemoglobin A1c) ELISA Kit, 96 tests
Catalog No. 501501095
Encompass_Preferred
Rat HbA1C (Glycosylated Hemoglobin/Hemoglobin A1c) ELISA Kit for detecting HbA1C in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 1.563-100ng/ml. Sensitivity: 0.938ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P01946. Target Synonyms: HbA1C; Glycosylated Hemoglobin; Hemoglobin A1c. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-1095 Supplier Biomatik Corporation Supplier No. EKF57935
Add to Cart
Add to Cart
Rat HbA1C (Glycosylated Hemoglobin/Hemoglobin A1c) ELISA Kit for detecting HbA1C in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 1.563-100ng/ml. Sensitivity: 0.938ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P01946. Target Synonyms: HbA1C; Glycosylated Hemoglobin; Hemoglobin A1c. Restrictions: For Research Use Only. Not for use in diagnostic procedures.