Learn More
Biomatik Corporation Rat IFN- (Interferon Beta) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKF57937
Encompass_Preferred
Rat IFN-β (Interferon Beta) ELISA Kit for detecting IFN-β in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 15.625-1000pg/ml. Sensitivity: 9.375pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P70499. Target Synonyms: IFN-β; IFNB1; Fibroblast interferon; IFBIFNB; IFF; IFN-beta; interferon beta. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-1097