Learn More
Biomatik Corporation Rat INS (Insulin) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKF57940
Encompass_Preferred
Rat INS (Insulin) ELISA Kit for detecting INS in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 78.125-5000pg/ml. Sensitivity: 46.875pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P01322. Target Synonyms: Insulin; INS. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-1100