Learn More
Biomatik Corporation Rat NF-B (Nuclear Factor Kappa B) ELISA Kit, 96 tests
Catalog No. 501501103
Encompass_Preferred
Rat NF-κB (Nuclear Factor Kappa B) ELISA Kit for detecting NF-κB in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.094ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: Q63369; Q7TQN4; D3Z9V1. Target Synonyms: NF-κB. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-1103 Supplier Biomatik Corporation Supplier No. EKF57947
Rat NF-κB (Nuclear Factor Kappa B) ELISA Kit for detecting NF-κB in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.094ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: Q63369; Q7TQN4; D3Z9V1. Target Synonyms: NF-κB. Restrictions: For Research Use Only. Not for use in diagnostic procedures.