Learn More
Biomatik Corporation Rat OC/BGP (Osteocalcin) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKF57948
Encompass_Preferred
Rat OC/BGP (Osteocalcin) ELISA Kit for detecting OC; BGP in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.313-20ng/ml. Sensitivity: 0.188ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P04640. Target Synonyms: OC; BGP. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-1104