Learn More
Biomatik Corporation Rat PCT (Procalcitonin) ELISA Kit, 96 tests
Catalog No. 501501107
Encompass_Preferred
Rat PCT (Procalcitonin) ELISA Kit for detecting PCT in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 15.625-1000pg/ml. Sensitivity: 9.375pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P01257. Target Synonyms: PCT; procalcitonin. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-1107 Supplier Biomatik Corporation Supplier No. EKF57951
Rat PCT (Procalcitonin) ELISA Kit for detecting PCT in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 15.625-1000pg/ml. Sensitivity: 9.375pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P01257. Target Synonyms: PCT; procalcitonin. Restrictions: For Research Use Only. Not for use in diagnostic procedures.