Learn More
Biomatik Corporation Rat Pg (Progesterone) ELISA Kit, 96 tests
Catalog No. 501501108
Encompass_Preferred
Rat Pg (Progesterone) ELISA Kit for detecting Pg in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.313-20ng/ml. Sensitivity: 0.188ng/ml. Format: 96-Well Plate. Assay Type: Competitive ELISA, Coated with Antigen. Assay Time: 3h, 30 min. Target Synonyms: PG; Progesterone. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-1108 Supplier Biomatik Corporation Supplier No. EKF57952
Rat Pg (Progesterone) ELISA Kit for detecting Pg in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.313-20ng/ml. Sensitivity: 0.188ng/ml. Format: 96-Well Plate. Assay Type: Competitive ELISA, Coated with Antigen. Assay Time: 3h, 30 min. Target Synonyms: PG; Progesterone. Restrictions: For Research Use Only. Not for use in diagnostic procedures.