Learn More
Biomatik Corporation Rat PRGN (progranulin) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKF57953
Encompass_Preferred
Rat PGRN (progranulin) ELISA Kit for detecting PGRN in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 62.5-4000pg/ml. Sensitivity: 37.5pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P23785. Target Synonyms: PGRN; Acrogranin; GEP; GP88; GRN; PCDGF; PEPI; Proepithelin. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-1109