Learn More
Biomatik Corporation Rat VEGFR-2/KDR (Vascular Endothelial Growth Factor Receptor 2) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKF57962
Encompass_Preferred
Rat VEGFR-2/KDR (Vascular Endothelial Growth Factor Receptor 2) ELISA Kit for detecting VEGFR-2; KDR in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.094ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: O08775. Target Synonyms: VEGFR2; KDR; VEGFR; VEGFR-2; CD309; Flk-1; CD309 antigen; Fetal liver kinase 1; fetal liver kinase-1; FLK-1; FLK1tyrosine kinase growth factor receptor; Kinase insert domain receptor; kinase insert domain receptor; a type III receptor tyrosine kinase; Protein-tyrosine kinase receptor flk-1; soluble VEGFR2; vascular endothelial growth factor receptor 2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-1118