Learn More
Biomatik Corporation Rat HCY (Homocysteine) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKF57965
Encompass_Preferred
Rat HCY (Homocysteine) ELISA Kit for detecting HCY in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 7.813-500pmol/ml. Sensitivity: 4.688pmol/ml. Format: 96-Well Plate. Assay Type: Competitive ELISA, Coated with Antigen. Assay Time: 3h, 30 min. Target Synonyms: HCY. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-1121