Learn More
Biomatik Corporation ST/5-HT (5-hydroxytryptamine) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKF57977
Encompass_Preferred
ST/5-HT (5-hydroxytryptamine) ELISA Kit for detecting ST; 5-HT in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Universal. Detection Range: 1.563-100ng/ml. Sensitivity: 0.938ng/ml. Format: 96-Well Plate. Assay Type: Competitive ELISA, Coated with Antigen. Assay Time: 3h, 30 min. Target Synonyms: ST; 5-HT; Serotonin; 5-Hydroxytryptamine. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-1126