Learn More
Biomatik Corporation FA/VB9 (Folic Acid/Vitamin B9) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKF57978
Encompass_Preferred
The FA/VB9 (Folic Acid/Vitamin B9) ELISA Kit allows for quantitative determination of FA/VB9 in Serum, plasma, tissue homogenates and other biological fluids. Species Reactivity: All species. Detection Range: 78.125-5000 pg/mL. Sensitivity: 69.748 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Competitive ELISA, Coated with Antigen. Restriction: For Research Use Only.
Catalog No. 50-150-1127